1-(4-Benzylpiperazin-1-yl)-2-(5-nitro-1H-indol-3-yl)ethane-1,2-dione

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Benzyl-4-[(5-nitro-1H-indol-3-yl)(oxo)acetyl]piperazine, 3-[(4-Benzylpiperazin-1-yl)(oxo)acetyl]-5-nitro-1H-indole

OR28200-50MG
APOSOR28200-50MGEA 109 EUR
APOSOR28200-50MG
1-(4-Benzylpiperazin-1-yl)-2-(5-nitro-1H-indol-3-yl)ethane-1,2-dione
1-(4-Benzylpiperazin-1-yl)-2-(5-nitro-1H-indol-3-yl)ethane-1,2-dione
Formel: C₂₁H₂₀N₄O₄
Molecular Weight: 392,408 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01935748
CAS-Nummer: 334973-49-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR