(Brommethyl)cyclopropan 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR28323-100G
OR28323-25G
OR28323-5G
APOSOR28323-100GEA
65.4
EUR
APOSOR28323-100G
APOSOR28323-25G
APOSOR28323-5G
(Brommethyl)cyclopropan 98%
(Brommethyl)cyclopropan
Formel:
C₄H₇Br Siedepunkt: 101…112 °C Dichte: 1,392 g/cm³ (25 °C) Flammpunkt: 41 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00001306 CAS-Nummer: 7051-34-5 EINECS: 230-331-8 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...