6-Amino-7-deazapurine hydrogen sulphate

Lieferant: Apollo Scientific
Achtung

OR2834-1G OR2834-5G OR2834-25G
APOSOR2834-1GEA 163 EUR
APOSOR2834-1G APOSOR2834-5G APOSOR2834-25G
6-Amino-7-deazapurine hydrogen sulphate
6-Amino-7-deazapurine hydrogen sulphate
Formel: C₆H₈N₄O₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD09787504
CAS-Nummer: 769951-32-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR