1,6-Naphthalindisulfonsäure-Dinatriumsalz 97%

Lieferant: Apollo Scientific
Achtung

OR2838-100G OR2838-500G OR2838-1KG
APOSOR2838-100GEA 21.9 EUR
APOSOR2838-100G APOSOR2838-500G APOSOR2838-1KG
1,6-Naphthalindisulfonsäure-Dinatriumsalz 97%
Disodium-1,6-naphthalenedisulphonate
Formel: C₁₀H₈O₆S₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00021522
CAS-Nummer: 1655-43-2
EINECS: 216-734-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR