3-Pyridinecarboxamide oxime

Lieferant: Apollo Scientific
Achtung

OR28384-100G OR28384-25G OR28384-5G
APOSOR28384-100GEA 0 EUR
APOSOR28384-100G APOSOR28384-25G APOSOR28384-5G
3-Pyridinecarboxamide oxime
3-Pyridinecarboxamide oxime
Formel: C₆H₇N₃O
Schmelzpunkt: 130…135 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00265955
CAS-Nummer: 1594-58-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR