6-Methyl-4-chinazolon 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Methylquinazolin-4-ol, 6-Methylquinazolin-4(3H)-one

OR2839-250MG
APOSOR2839-250MGEA 21.9 EUR
APOSOR2839-250MG
6-Methyl-4-chinazolon 97%
6-Methyl-4-chinazolon
Formel: C₉H₈N₂O
Molecular Weight: 160,1726 g/mol
Schmelzpunkt: 238…240 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00673185
CAS-Nummer: 19181-53-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR