5-Methylamino[1,3,4]thiadiazole-2-thiol 95%

Lieferant: Apollo Scientific

OR28426-1G
APOSOR28426-1GEA 644 EUR
APOSOR28426-1G
5-Methylamino[1,3,4]thiadiazole-2-thiol 95%
5-Methylamino[1,3,4]thiadiazole-2-thiol
Formel: C₃H₅N₃S₂
Molecular Weight: 147,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00089421
CAS-Nummer: 27386-01-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR