7-Chloro-1-benzothiophene-2-carboxylic acid 90%

Lieferant: Apollo Scientific
Achtung

OR2843-1G
APOSOR2843-1GEA 49.3 EUR
APOSOR2843-1G
7-Chloro-1-benzothiophene-2-carboxylic acid 90%
7-Chloro-1-benzothiophene-2-carboxylic acid
Formel: C₉H₅ClO₂S
Molecular Weight: 212,6500 g/mol
Schmelzpunkt: 289…291 °C
Lagertemperatur: Kühlschrank
MDL: MFCD07377072
CAS-Nummer: 90407-16-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR