5-Chloro-1H-indole-3-carbaldehyde

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Chloroindole-3-carboxaldehyde

OR28433-1G
APOSOR28433-1GEA 69.6 EUR
APOSOR28433-1G
5-Chloro-1H-indole-3-carbaldehyde
5-Chloro-1H-indole-3-carbaldehyde
Formel: C₉H₆ClNO
Molecular Weight: 179,61 g/mol
Schmelzpunkt: 211…214 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00175291
CAS-Nummer: 827-01-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR