N-Hydroxysuccinimid 99%
Lieferant: Apollo Scientific
|
Achtung
|
OR28434-100G
OR28434-1KG
OR28434-250G
APOSOR28434-100GEA
21.9
EUR
APOSOR28434-100G
APOSOR28434-1KG
APOSOR28434-250G
N-Hydroxysuccinimid 99%
N-Hydroxysuccinimid
Formel:
C₄H₅NO₃ Schmelzpunkt: 94…99 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005516 CAS-Nummer: 6066-82-6 EINECS: 228-001-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...