N-Hydroxysuccinimid 99%

Lieferant: Apollo Scientific
Achtung

OR28434-100G OR28434-1KG OR28434-250G
APOSOR28434-100GEA 21.9 EUR
APOSOR28434-100G APOSOR28434-1KG APOSOR28434-250G
N-Hydroxysuccinimid 99%
N-Hydroxysuccinimid
Formel: C₄H₅NO₃
Schmelzpunkt: 94…99 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005516
CAS-Nummer: 6066-82-6
EINECS: 228-001-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR