1-Naphthylacetyl chloride
Lieferant: Apollo Scientific
|
Gefahr
|
OR28451-1G
APOSOR28451-1GEA
87
EUR
APOSOR28451-1G
1-Naphthylacetyl chloride
1-Naphthylacetyl chloride
Formel:
C₁₂H₉ClO Molecular Weight: 204,66 g/mol Siedepunkt: 130 °C/0,6mmHg Dichte: 1,23 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00175852 CAS-Nummer: 5121-00-6 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...