Benzo[b]thiophene-3-acetonitrile

Lieferant: Apollo Scientific

OR28492-1G
APOSOR28492-1GEA 161 EUR
APOSOR28492-1G
Benzo[b]thiophene-3-acetonitrile
Benzo[b]thiophene-3-acetonitrile
Formel: C₁₀H₇NS
Molecular Weight: 173,24 g/mol
Schmelzpunkt: 62…66 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00027389
CAS-Nummer: 3216-48-6
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR