1,4-Bis(4-phenoxybenzoyl)benzene

Lieferant: Apollo Scientific
Achtung

Synonyme: Phenylene-1,4-bis[(4-phenoxyphenyl)methanone]

OR2850-1G
APOSOR2850-1GEA 140 EUR
APOSOR2850-1G
1,4-Bis(4-phenoxybenzoyl)benzene
1,4-Bis(4-phenoxybenzoyl)benzene
Formel: C₃₂H₂₂O₄
Molecular Weight: 470,515 g/mol
Schmelzpunkt: 216…218 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00319174
CAS-Nummer: 54299-17-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR