3-{[2-Phenylvinyl]thio}propanoic acid
Lieferant: Apollo Scientific
OR28509-1G
APOSOR28509-1GEA
72.5
EUR
APOSOR28509-1G
3-{[2-Phenylvinyl]thio}propanoic acid
3-{[2-Phenylvinyl]thio}propanoic acid
Formel:
C₁₁H₁₂O₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD00068080 CAS-Nummer: 175205-21-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...