2,3-Dichlorisonicotinsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,3-Dichloropyridine-4-carboxylic acid
OR2859-1G
OR2859-250MG
APOSOR2859-1GEA
24.8
EUR
APOSOR2859-1G
APOSOR2859-250MG
2,3-Dichlorisonicotinsäure
2,3-Dichlorisonicotinsäure
Formel:
C₆H₃Cl₂NO₂ Molecular Weight: 192 g/mol Schmelzpunkt: 187 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01861977 CAS-Nummer: 184416-84-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...