6-Nitro-1H-benzimidazolnitrat

Lieferant: Apollo Scientific

OR28596-250G
APOSOR28596-250GEA 248 EUR
APOSOR28596-250G
6-Nitro-1H-benzimidazolnitrat
6-Nitro-1H-benzimidazolnitrat
Formel: C₇H₅N₃O₂
Molecular Weight: 226,15 g/mol
Schmelzpunkt: 222 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013157
CAS-Nummer: 27896-84-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR