(3,4-Dimethoxyphenyl)essigsäureethylester 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl-(3,4-dimethoxyphenyl)acetat

OR28612-100G
APOSOR28612-100GEA 194 EUR
APOSOR28612-100G
(3,4-Dimethoxyphenyl)essigsäureethylester 98%
(3,4-Dimethoxyphenyl)essigsäureethylester
Formel: C₁₂H₁₆O₄
Molecular Weight: 224,26 g/mol
Siedepunkt: 159…160 °C/4mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00017272
CAS-Nummer: 18066-68-7
EINECS: 241-974-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR