1,8-Dibromoctan 98%
Lieferant: Apollo Scientific
Synonyme:
Octamethylenbromid
OR28631-100G
APOSOR28631-100GEA
92.8
EUR
APOSOR28631-100G
1,8-Dibromoctan 98%
1,8-Dibromoctan
Formel:
BrH₂C(CH₂)₆CH₂Br Molecular Weight: 272,02 g/mol Siedepunkt: 270…272 °C Schmelzpunkt: 15…16 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000277 CAS-Nummer: 4549-32-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...