6-Phenyl-2-pyridincarbonsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Phenylpicolinic acid, 6-Phenylpyridine-2-carboxylic acid

OR2864-250MG
APOSOR2864-250MGEA 21.9 EUR
APOSOR2864-250MG
6-Phenyl-2-pyridincarbonsäure
6-Phenylpicolinic acid
Formel: C₁₂H₉NO₂
Molecular Weight: 199,2054 g/mol
Schmelzpunkt: 95…100 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04114206
CAS-Nummer: 39774-28-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR