2-Isopropoxyphenol 97%

Lieferant: Apollo Scientific

OR28649-25G
APOSOR28649-25GEA 115 EUR
APOSOR28649-25G
2-Isopropoxyphenol 97%
2-Isopropoxyphenol
Formel: C₉H₁₂O₂
Molecular Weight: 152,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00002184
CAS-Nummer: 4812-20-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR