4-Chlorthiobenzamid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Chlorthiobenzoesäureamid
OR28662-1G
APOSOR28662-1GEA
21.9
EUR
APOSOR28662-1G
4-Chlorthiobenzamid
4-Chlorthiobenzamid
Formel:
C₇H₆ClNS Molecular Weight: 171,65 g/mol Schmelzpunkt: 127…129 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00040956 CAS-Nummer: 2521-24-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...