Methyl-2-(aminosulphonyl)benzoate 98%

Lieferant: Apollo Scientific

OR28690-500G
APOSOR28690-500GEA 0 EUR
APOSOR28690-500G
Methyl-2-(aminosulphonyl)benzoate 98%
Methyl-2-(aminosulphonyl)benzoate
Formel: C₈H₉NO₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD00009808
CAS-Nummer: 57683-71-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR