3,4-Dihydro-4-oxo-1,2,3-benzotriazine

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Ketobenzotriazine, 1,2,3-Benzotriazine-4(3H)-one, Benzo[d][1,2,3]triazin-4(3H)-one

OR28692-10G
APOSOR28692-10GEA 36.4 EUR
APOSOR28692-10G
3,4-Dihydro-4-oxo-1,2,3-benzotriazine
3,4-Dihydro-4-oxo-1,2,3-benzotriazine
Formel: C₇H₅N₃O
Molecular Weight: 147,14 g/mol
Schmelzpunkt: 216…224 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052387
CAS-Nummer: 90-16-4
EINECS: 201-971-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR