Dimethyl-2-aminoterephthalat
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Dimethyl aminoterephthalate, 2-Aminodimethyl terephthalate
OR28713-5G
APOSOR28713-5GEA
21.9
EUR
APOSOR28713-5G
Dimethyl-2-aminoterephthalat
Dimethyl-2-aminoterephthalat
Formel:
C₁₀H₁₁NO₄ Molecular Weight: 209,2 g/mol Schmelzpunkt: 127…134 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008427 CAS-Nummer: 5372-81-6 EINECS: 226-364-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...