4-(Imidazo[1,2-a]pyrid-2-yl)aniline

Lieferant: Apollo Scientific
Achtung

OR2872-1G
APOSOR2872-1GEA 109 EUR
APOSOR2872-1G
4-(Imidazo[1,2-a]pyrid-2-yl)aniline
4-(Imidazo[1,2-a]pyrid-2-yl)aniline
Formel: C₁₃H₁₁N₃
Molecular Weight: 209,252 g/mol
Schmelzpunkt: 230 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00107394
CAS-Nummer: 139705-74-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR