6-Chlor-m-toluylsäure

Lieferant: Apollo Scientific
Achtung

OR2875-1G
APOSOR2875-1GEA 21.9 EUR
APOSOR2875-1G
6-Chlor-m-toluylsäure
6-Chlor-m-toluylsäure
Formel: C₈H₇ClO₂
Molecular Weight: 170.6 g/mol
Schmelzpunkt: 148…155 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00045798
CAS-Nummer: 6342-60-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR