Diethyl(2-cyanethyl)malonat
Lieferant: Apollo Scientific
Synonyme:
Diethyl 2-(2-cyanoethyl)malonate
OR28758-25G
APOSOR28758-25GEA
95.7
EUR
APOSOR28758-25G
Diethyl(2-cyanethyl)malonat
Diethyl(2-cyanethyl)malonat
Formel:
C₁₀H₁₅NO₄ Molecular Weight: 213,23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001966 CAS-Nummer: 17216-62-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...