1-Adamantancarbonsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Adamantanecarboxylic acid, Tricyclo[3.3.1.1(3,7)]decane-1-carboxylic acid
OR28762-100G
OR28762-25G
OR28762-500G
APOSOR28762-100GEA
40.6
EUR
APOSOR28762-100G
APOSOR28762-25G
APOSOR28762-500G
1-Adamantancarbonsäure 98%
1-Adamantancarbonsäure
Formel:
C₁₁H₁₆O₂ Schmelzpunkt: 172…176 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00074720 CAS-Nummer: 828-51-3 EINECS: 212-584-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...