Methyl-3-amino-4-methyl-2-thiophencarboxylat
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 3-amino-4-methyl-2-thiophenecarboxylate, 2-Thiophenecarboxylic acid, 3-amino-4-methyl-, methyl ester, Methyl 3-amino-4-methylthiophene-2-carboxylate
OR28776-25G
APOSOR28776-25GEA
21.9
EUR
APOSOR28776-25G
Methyl-3-amino-4-methyl-2-thiophencarboxylat
Methyl-3-amino-4-methylthiophene-2-carboxylate
Formel:
C₇H₉NO₂S Molecular Weight: 171.21 g/mol Schmelzpunkt: 84…88 °C Flammpunkt: >100 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051822 CAS-Nummer: 85006-31-1 EINECS: 285-060-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...