6-Hydroxy-m-toluylsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Methylsalicylic acid, 2,5-Cresotic acid
OR28783-100G
OR28783-25G
APOSOR28783-100GEA
85.7
EUR
APOSOR28783-100G
APOSOR28783-25G
6-Hydroxy-m-toluylsäure
6-Hydroxy-m-toluylsäure
Formel:
C₈H₈O₃ Molecular Weight: 152.15 g/mol Siedepunkt: 313 Schmelzpunkt: 150…154 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002461 CAS-Nummer: 89-56-5 EINECS: 201-918-6 Merck Index: 12,02650 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...