2-Methyl-1H-imidazole-4-carbaldehyde

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Methyl-1H-imidazole-5-carbaldehyde

OR28795-5G
APOSOR28795-5GEA 313 EUR
APOSOR28795-5G
2-Methyl-1H-imidazole-4-carbaldehyde
2-Methyl-1H-imidazole-4-carbaldehyde
Formel: C₅H₆N₂O
Molecular Weight: 110.12 g/mol
Schmelzpunkt: 165 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00173725
CAS-Nummer: 35034-22-1
UN: 3259
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR