2-Amino-5-methyl-1,3,4-thiadiazol

Lieferant: Apollo Scientific
Achtung

OR28803-5G
APOSOR28803-5GEA 21.9 EUR
APOSOR28803-5G
2-Amino-5-methyl-1,3,4-thiadiazol
2-Amino-5-methyl-1,3,4-thiadiazole
Formel: C₃H₅N₃S
Molecular Weight: 115,16 g/mol
Schmelzpunkt: 223…228 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003110
CAS-Nummer: 108-33-8
EINECS: 203-573-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR