(R)-(-)-Glycidylbutyrat
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
(-)-[(2R)-Oxiran-2-yl]methyl butanoate, (R)-Oxiran-2-ylmethyl butyrate
OR28822-100G
OR28822-25G
OR28822-5G
APOSOR28822-100GEA
34.8
EUR
APOSOR28822-100G
APOSOR28822-25G
APOSOR28822-5G
(R)-(-)-Glycidylbutyrat
(R)-(-)-Glycidylbutyrat
Formel:
C₇H₁₂O₃ Molecular Weight: 144.17 g/mol Siedepunkt: 197 °C Dichte: 1.032 g/cm³ (25 °C) Flammpunkt: 84 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00075120 CAS-Nummer: 60456-26-0 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...