6-Aminopenicillansäure 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: (2S,5R,6R)-6-Amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid 6-APA 6-Amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid 6-amino-2,2-dimethyl-5-oxoperhydroazeto[2,1-b][1,3]thiazole-3-carboxylic acid, (2S,5R,6R)-6-Amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, (+)-6-Aminopenicillanic acid, (2S,5R,6R)-(+)-6-Amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid

OR28860-500G
APOSOR28860-500GEA 97.3 EUR
APOSOR28860-500G
6-Aminopenicillansäure 97%
6-Aminopenicillanic acid
Formel: C₈H₁₂N₂O₃S
Molecular Weight: 216.26 g/mol
Schmelzpunkt: 198…200 °C (Zers.)
Lagertemperatur: Kühlschrank
MDL: MFCD00005176
CAS-Nummer: 551-16-6
EINECS: 208-993-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR