4-Hydroxybenzylaceton
Lieferant: Apollo Scientific
|
Achtung
|
OR28913-1G
APOSOR28913-1GEA
21.9
EUR
APOSOR28913-1G
4-Hydroxybenzylaceton
4-Hydroxybenzylaceton
Formel:
C₁₀H₁₂O₂ Schmelzpunkt: 82…83 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002394 CAS-Nummer: 5471-51-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...