5-(Chloromethyl)quinolin-8-ol hydrochloride

Lieferant: Apollo Scientific

Synonyme: 5-(Chloromethyl)-8-quinolinol Hydrochloride

OR2892-1G
APOSOR2892-1GEA 132 EUR
APOSOR2892-1G
5-(Chloromethyl)quinolin-8-ol hydrochloride
5-(Chloromethyl)quinolin-8-ol hydrochloride
Formel: C₁₀H₉Cl₂NO
Molecular Weight: 230,1 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01790514
CAS-Nummer: 4053-45-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR