2-Methyl-1H-imidazole-4-carbothioamide

Lieferant: Apollo Scientific
Achtung

OR28983-1G
APOSOR28983-1GEA 79.9 EUR
APOSOR28983-1G
2-Methyl-1H-imidazole-4-carbothioamide
2-Methyl-1H-imidazole-4-carbothioamide
Formel: C₅H₇N₃S
Molecular Weight: 141,194 g/mol
Schmelzpunkt: 194…197 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03659724
CAS-Nummer: 129486-91-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR