4-(1,2,3-Thiadiazol-4-yl)aniline

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(4-Aminophenyl)-1,2,3-thiadiazole

OR29027-250MG
APOSOR29027-250MGEA 109 EUR
APOSOR29027-250MG
4-(1,2,3-Thiadiazol-4-yl)aniline
4-(1,2,3-Thiadiazol-4-yl)aniline
Formel: C₈H₇N₃S
Molecular Weight: 177,226 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00052104
CAS-Nummer: 121180-51-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR