3-(Chloromethyl)-5-(1,2,3-thiadiazol-4-yl)-1,2,4-oxadiazole
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-[3-(Chloromethyl)-1,2,4-oxadiazol-5-yl]-1,2,3-thiadiazole
OR29042-250MG
APOSOR29042-250MGEA
163
EUR
APOSOR29042-250MG
3-(Chloromethyl)-5-(1,2,3-thiadiazol-4-yl)-1,2,4-oxadiazole
3-(Chloromethyl)-5-(1,2,3-thiadiazol-4-yl)-1,2,4-oxadiazole
Formel:
C₅H₃ClN₄OS Molecular Weight: 202,622 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01313801 CAS-Nummer: 253586-59-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...