3-(1,3-Oxazol-5-yl)anilin
Lieferant: Apollo Scientific
|
Achtung
|
OR29046-1G
OR29046-250MG
APOSOR29046-1GEA
0
EUR
APOSOR29046-1G
APOSOR29046-250MG
3-(1,3-Oxazol-5-yl)anilin
3-(1,3-Oxazol-5-yl)anilin
Formel:
C₉H₈N₂O Schmelzpunkt: 93…95 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052191 CAS-Nummer: 157837-31-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...