3,4-Thiophendicarbonsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: Thiophene-3,4-dicarboxylic acid

OR2907-25G
APOSOR2907-25GEA 127 EUR
APOSOR2907-25G
3,4-Thiophendicarbonsäure
3,4-Thiophendicarbonsäure
Formel: C₆H₄O₄S
Molecular Weight: 172.15 g/mol
Schmelzpunkt: 227…231 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00067079
CAS-Nummer: 4282-29-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR