2-Hydroxy-5-nitro-4-picolin 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Methyl-5-nitro-2-pyridon, 2-Hydroxy-4-methyl-5-nitropyridin
OR2913-100G
APOSOR2913-100GEA
232
EUR
APOSOR2913-100G
2-Hydroxy-5-nitro-4-picolin 95+%
2-Hydroxy-5-nitro-4-picolin
Formel:
C₆H₆N₂O₃ Molecular Weight: 154,13 g/mol Schmelzpunkt: 186…190 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010690 CAS-Nummer: 21901-41-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...