5-(5-Bromothien-2-yl)isoxazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-(5-Bromothien-2-yl)-1,2-oxazole, 2-Bromo-5-(isoxazol-5-yl)thiophene

OR29131-1G
APOSOR29131-1GEA 65.4 EUR
APOSOR29131-1G
5-(5-Bromothien-2-yl)isoxazole
5-(5-Bromothien-2-yl)isoxazole
Formel: C₇H₄BrNOS
Molecular Weight: 230,082 g/mol
Schmelzpunkt: 66…67 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03618406
CAS-Nummer: 138716-31-1
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR