3'-Cyanoacetanilide

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(Acetylamino)benzonitrile, N-(3-Cyanophenyl)acetamide

OR29135-25G
APOSOR29135-25GEA 993 EUR
APOSOR29135-25G
3'-Cyanoacetanilide
3'-Cyanoacetanilide
Formel: C₉H₈N₂O
Molecular Weight: 160,173 g/mol
Schmelzpunkt: 130…132 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01047361
CAS-Nummer: 58202-84-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR