2-Chlor-4-methyl-3-nitropyridin 98+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Chlor-3-nitro-4-picolin
OR2914-5G
OR2914-25G
OR2914-100G
APOSOR2914-5GEA
21.9
EUR
APOSOR2914-5G
APOSOR2914-25G
APOSOR2914-100G
2-Chlor-4-methyl-3-nitropyridin 98+%
2-Chloro-4-methyl-3-nitropyridine
Formel:
C₆H₅ClN₂O₂ Molecular Weight: 172.57 g/mol Schmelzpunkt: 51…53 °C Flammpunkt: 113 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012347 CAS-Nummer: 23056-39-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...