2,3-Dihydro-6-phenylimidazo[2,1-b]thiazole-5-carboxaldehyde
Lieferant: Apollo Scientific
Synonyme:
6-phenyl-2,3-dihydroimidazo[2,1-b][1,3]thiazole-5-carboxaldehyde
OR29177-1G
APOSOR29177-1GEA
145
EUR
APOSOR29177-1G
2,3-Dihydro-6-phenylimidazo[2,1-b]thiazole-5-carboxaldehyde
2,3-Dihydro-6-phenylimidazo[2,1-b]thiazole-5-carboxaldehyde
Formel:
C₁₂H₁₀N₂OS Molecular Weight: 230,29 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00268936 CAS-Nummer: 75224-64-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...