Tri(2-thienyl)phosphine

Lieferant: Apollo Scientific
Achtung

Synonyme: Tri(thiophen-2-yl)phosphine

OR29178-1G
APOSOR29178-1GEA 0 EUR
APOSOR29178-1G
Tri(2-thienyl)phosphine
Tri(2-thienyl)phosphine
Formel: C₁₂H₉PS₃
Molecular Weight: 280,36 g/mol
Schmelzpunkt: 28…30 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00015471
CAS-Nummer: 24171-89-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR