2-(1-Methylhydrazino)quinoxaline
Lieferant: Apollo Scientific
|
Achtung
|
OR29181-250MG
APOSOR29181-250MGEA
21.9
EUR
APOSOR29181-250MG
2-(1-Methylhydrazino)quinoxaline
2-(1-Methylhydrazino)quinoxaline
Formel:
C₉H₁₀N₄ Schmelzpunkt: 108…110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00085138 CAS-Nummer: 16621-55-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...