4-Aminopyrimidin-5-carbosäurenitril
Lieferant: Apollo Scientific
OR29186-25G
APOSOR29186-25GEA
183
EUR
APOSOR29186-25G
4-Aminopyrimidin-5-carbosäurenitril
4-Aminopyrimidin-5-carbosäurenitril
Formel:
C₅H₄N₄ Molecular Weight: 120,11 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00054552 CAS-Nummer: 16357-69-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...